Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC034259.1 | - | 3 - 4 | 395 | 4238589 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |