Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080554.2 | NP_542121.2 | 3 - 4 | 475 | 71 | |
GenBank mRNA | AK005335.1 | - | 3 - 4 | 453 | 71 | |
AK011026.1 | - | 3 - 4 | 452 | 71 | ||
AK032992.1 | - | 3 - 4 | 453 | 71 | ||
AK145587.1 | - | 3 - 4 | 452 | 71 | ||
AK148284.1 | - | 476 | 71 | |||
AK220321.1 | - | 3 - 4 | 416 | 71 | ||
BC019112.1 | - | 3 - 4 | 486 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |