Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ak8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033874.2 | NP_001029046.2 | 3 - 4 | 243 | 87 | |
GenBank mRNA | BC100481.1 | - | 3 - 4 | 240 | 30917849 | 87 |
BC109352.2 | - | 3 - 4 | 230 | 40057006 | 87 | |
BY100469.1 | - | 3 - 4 | 244 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |