Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145528.3 | NP_663503.1 | 4 - 5 | 633 | 148 | |
GenBank mRNA | AK149672.1 | - | 4 - 5 | 592 | 148 | |
AK171206.1 | - | 4 - 5 | 619 | 148 | ||
AK171440.1 | - | 4 - 5 | 610 | 148 | ||
BC016669.1 | - | 4 - 5 | 617 | 2648797 | 148 | |
BC023673.1 | - | 4 - 5 | 583 | 5343236 | 148 | |
BC023712.1 | - | 4 - 5 | 606 | 5346303 | 148 | |
BC033419.1 | - | 4 - 5 | 584 | 5359944 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |