Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244198.1 | NP_001231127.1 | 5 - 6 | 472 | 80 | |
NM_001244200.1 | NP_001231129.1 | 6 - 7 | 684 | 80 | ||
NM_013627.5 | NP_038655.1 | 5 - 6 | 1028 | 80 | ||
GenBank mRNA | AK032616.1 | - | 5 - 6 | 473 | 80 | |
BC011272.1 | - | 5 - 6 | 716 | 4008490 | 80 | |
X63963.1 | - | 5 - 6 | 349 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |