Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144824.2 | NP_659073.1 | 7 - 8 | 1390 | 75 | |
GenBank mRNA | AK077570.1 | - | 7 - 8 | 1294 | 75 | |
AK145219.1 | - | 7 - 8 | 1317 | 75 | ||
AK169271.1 | - | 6 - 7 | 1403 | 75 | ||
BC021790.1 | - | 7 - 8 | 1390 | 4220923 | 75 | |
BC069868.1 | - | 8 - 9 | 1434 | 5101220 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |