Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028360.2 | NP_082636.3 | 7 - 8 | 689 | 130 | |
NM_029704.2 | NP_083980.2 | 6 - 7 | 650 | 130 | ||
GenBank mRNA | AK034075.1 | - | 7 - 8 | 679 | 130 | |
BC026866.1 | - | 5 - 6 | 333 | 130 | ||
BC037111.1 | - | 6 - 7 | 413 | 5370464 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |