Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029911.4 | NP_084187.2 | 4 - 5 | 764 | 82 | |
GenBank mRNA | AK031904.1 | - | 4 - 5 | 695 | 82 | |
AK036066.1 | - | 4 - 5 | 1038 | 82 | ||
AK082153.1 | - | 4 - 5 | 765 | 82 | ||
BC132487.1 | - | 4 - 5 | 706 | 40130764 | 82 | |
BC137869.1 | - | 4 - 5 | 706 | 8860888 | 82 | |
DQ185134.1 | - | 4 - 5 | 661 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |