Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008569.2 | NP_032595.2 | 37 - 38 | 4779 | 130 | |
GenBank mRNA | AK034858.1 | - | 5 - 6 | 310 | 130 | |
AK090134.1 | - | 37 - 38 | 4761 | 130 | ||
BC139002.1 | - | 37 - 38 | 4760 | 8862024 | 130 | |
BC145492.1 | - | 37 - 38 | 4760 | 9054055 | 130 | |
X80169.1 | - | 37 - 38 | 4764 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |