Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021398.3 | NP_067373.2 | 11 - 12 | 1663 | 74 | |
GenBank mRNA | AF188622.1 | - | 11 - 12 | 1658 | 74 | |
BC089603.1 | - | 10 - 11 | 1363 | 6758493 | 74 | |
BU516072.1 | - | 5 - 6 | 699 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |