Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081349.2 | NP_001074818.2 | 2 - 3 | 437 | 75 | |
NM_001083809.1 | NP_001077278.1 | 1 - 2 | 376 | 75 | ||
NM_024497.1 | NP_078773.1 | 2 - 3 | 335 | 75 | ||
GenBank mRNA | AB103034.1 | - | 2 - 3 | 406 | 75 | |
AK011417.1 | - | 2 - 3 | 435 | 75 | ||
AK031215.1 | - | 1 - 2 | 379 | 75 | ||
AK046758.1 | - | 2 - 3 | 429 | 75 | ||
AK165374.1 | - | 2 - 3 | 429 | 75 | ||
BC053747.1 | - | 2 - 3 | 234 | 6330614 | 75 | |
BX522717.1 | - | 2 - 3 | 297 | 75 | ||
CJ186653.1 | - | 2 - 3 | 336 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |