Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085448.1 | NP_001078917.1 | 16 - 17 | 3058 | 100 | |
NM_001085453.1 | NP_001078922.1 | 15 - 16 | 2810 | 100 | ||
GenBank mRNA | AB093237.1 | - | 17 - 18 | 2892 | 100 | |
AK161790.1 | - | 17 - 18 | 2952 | 100 | ||
Z17804.1 | - | 2953 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |