Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026044.3 | NP_080320.1 | 4 - 5 | 522 | 74 | |
GenBank mRNA | AK013297.1 | - | 4 - 5 | 502 | 74 | |
AK078448.1 | - | 4 - 5 | 485 | 74 | ||
AK087998.1 | - | 3 - 4 | 397 | 74 | ||
AK152000.1 | - | 3 - 4 | 435 | 74 | ||
AK153537.1 | - | 3 - 4 | 435 | 74 | ||
BC056962.1 | - | 2 - 3 | 1172 | 74 | ||
BC086486.1 | - | 4 - 5 | 524 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |