Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013830.2 | NP_038858.2 | 13 - 14 | 2900 | 124 | |
GenBank mRNA | AB516304.1 | - | 13 - 14 | 2752 | 124 | |
AF033663.1 | - | 12 - 13 | 2190 | 124 | ||
AF283466.1 | - | 13 - 14 | 2898 | 124 | ||
AK076020.1 | - | 5 - 6 | 710 | 124 | ||
AK083926.1 | - | 13 - 14 | 2784 | 124 | ||
BC003769.1 | - | 11 - 12 | 1418 | 124 | ||
BC042521.1 | - | 14 - 15 | 2182 | 124 | ||
BC080864.1 | - | 13 - 14 | 2871 | 124 | ||
BC141272.1 | - | 13 - 14 | 2826 | 9055884 | 124 | |
BC141273.1 | - | 13 - 14 | 2826 | 9055885 | 124 | |
U48737.1 | - | 11 - 12 | 1457 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |