Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146122.3 | NP_666234.3 | 20 - 21 | 1832 | 70 | |
GenBank mRNA | AK136192.1 | - | 13 - 14 | 70 | ||
AK137530.1 | - | 19 - 20 | 1760 | 70 | ||
AK220407.1 | - | 18 - 19 | 1579 | 70 | ||
BC027786.1 | - | 20 - 21 | 1795 | 5346095 | 70 | |
CX563166.1 | - | 2 - 3 | 206 | 70 | ||
DQ448594.1 | - | 20 - 21 | 1577 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |