Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013515.2 | NP_038543.1 | 5 - 6 | 614 | 82 | |
GenBank mRNA | AF093620.1 | - | 5 - 6 | 565 | 82 | |
AK077435.1 | - | 5 - 6 | 582 | 82 | ||
AK139242.1 | - | 5 - 6 | 582 | 82 | ||
AK143724.1 | - | 5 - 6 | 582 | 82 | ||
AK148975.1 | - | 5 - 6 | 583 | 82 | ||
AK149689.1 | - | 5 - 6 | 582 | 82 | ||
AK149821.1 | - | 5 - 6 | 582 | 82 | ||
AK149991.1 | - | 5 - 6 | 582 | 82 | ||
AK151129.1 | - | 5 - 6 | 581 | 82 | ||
U17297.1 | - | 5 - 6 | 614 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |