Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145522.4 | NP_663497.2 | 4 - 5 | 531 | 106 | |
GenBank mRNA | AK035343.1 | - | 4 - 5 | 411 | 106 | |
AK153022.1 | - | 4 - 5 | 435 | 106 | ||
BC019800.1 | - | 4 - 5 | 407 | 5150243 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |