Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011706.2 | NP_035836.2 | 2 - 3 | 273 | 76 | |
GenBank mRNA | AK134052.1 | - | 1 - 2 | 586 | 76 | |
AK171222.1 | - | 1 - 2 | 196 | 76 | ||
BB631765.1 | - | 2 - 3 | 273 | 76 | ||
BC005415.1 | - | 1 - 2 | 220 | 3487527 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus 1700 Probe spans exons |