Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154760.1 | - | 13 - 14 | 1623 | 71 | |
AK161339.1 | - | 8 - 9 | 913 | 71 | ||
BC065140.1 | - | 13 - 14 | 1495 | 71 | ||
BC086671.1 | - | 13 - 14 | 1500 | 30635559 | 71 | |
D38557.1 | - | 13 - 14 | 1570 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |