Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027163.4 | NP_081439.1 | 1 - 2 | 58 | 136 | |
GenBank mRNA | AK009787.1 | - | 1 - 2 | 70 | 136 | |
AY071842.1 | - | 1 - 2 | 43 | 136 | ||
BC119517.1 | - | 1 - 2 | 58 | 40055701 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |