Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029758.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029758.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029743.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029743.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029743.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029754.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029754.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029755.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029755.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029759.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029759.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029731.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029731.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029731.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029744.mm.1 |
NM_001289521.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029744.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029744.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029760.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029760.mm.1 |
TC0200003473.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029757.mm.1 |
NM_015753.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029757.mm.1 |
TC0200003473.mm.1 |
MTA TC |