Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139308.2 | NP_647469.2 | 4 - 5 | 1026 | 83 | |
GenBank mRNA | AF244543.1 | - | 3 - 4 | 383 | 83 | |
AK083125.1 | - | 3 - 4 | 781 | 83 | ||
AK143699.1 | - | 4 - 5 | 1027 | 83 | ||
BC017524.1 | - | 4 - 5 | 1016 | 4527790 | 83 | |
BC080747.1 | - | 4 - 5 | 997 | 4237000 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |