Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190817.1 | NP_001177746.1 | 3 - 4 | 683 | 151 | |
NM_007869.3 | NP_031895.1 | 3 - 4 | 683 | 151 | ||
GenBank mRNA | AK133582.1 | - | 3 - 4 | 667 | 151 | |
AK144228.1 | - | 3 - 4 | 639 | 151 | ||
BC080300.1 | - | 3 - 4 | 664 | 151 | ||
L16953.1 | - | 3 - 4 | 360 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |