Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007578.2 | NP_001007579.1 | 2 - 3 | 186 | 83 | |
GenBank mRNA | AK148507.1 | - | 2 - 3 | 185 | 83 | |
AK160766.1 | - | 2 - 3 | 187 | 83 | ||
BC027264.1 | - | 2 - 3 | 179 | 3492516 | 83 | |
BY777182.1 | - | 2 - 3 | 166 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |