Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 8 - 9 | 1049 | 65 | |
GenBank mRNA | AK029123.1 | - | 9 - 10 | 1447 | 65 | |
AK041436.1 | - | 9 - 10 | 1417 | 65 | ||
AK052264.1 | - | 9 - 10 | 1436 | 65 | ||
AK122426.1 | - | 9 - 10 | 1139 | 65 | ||
BC058971.1 | - | 8 - 9 | 991 | 6832507 | 65 | |
BC116262.1 | - | 8 - 9 | 1016 | 65 | ||
BC116263.1 | - | 8 - 9 | 1016 | 65 | ||
BC129961.1 | - | 8 - 9 | 1016 | 65 | ||
BC129962.1 | - | 8 - 9 | 1016 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |