Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170978.1 | NP_001164449.1 | 8 - 9 | 728 | 75 | |
NM_172961.3 | NP_766549.2 | 8 - 9 | 728 | 75 | ||
GenBank mRNA | AK036128.1 | - | 8 - 9 | 673 | 75 | |
AK132282.1 | - | 8 - 9 | 677 | 75 | ||
AK165348.1 | - | 8 - 9 | 678 | 75 | ||
BC058079.1 | - | 8 - 9 | 668 | 6826741 | 75 | |
BC058521.1 | - | 8 - 9 | 668 | 6825487 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |