Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159317.1 | NP_001152789.1 | 5 - 6 | 884 | 95 | |
NM_001159318.1 | NP_001152790.1 | 5 - 6 | 884 | 95 | ||
NM_008364.2 | NP_032390.1 | 5 - 6 | 884 | 95 | ||
NM_134103.2 | NP_598864.1 | 5 - 6 | 884 | 95 | ||
GenBank mRNA | AK033972.1 | - | 5 - 6 | 833 | 95 | |
AK034269.1 | - | 5 - 6 | 848 | 95 | ||
AK039582.1 | - | 5 - 6 | 836 | 95 | ||
AK045686.1 | - | 5 - 6 | 833 | 95 | ||
AK083209.1 | - | 5 - 6 | 881 | 95 | ||
AK136782.1 | - | 5 - 6 | 835 | 95 | ||
AK149467.1 | - | 5 - 6 | 833 | 95 | ||
BC021159.1 | - | 5 - 6 | 833 | 4161899 | 95 | |
X85999.1 | - | 5 - 6 | 835 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |