Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205321.1 | NP_001192250.1 | 20 - 21 | 3577 | 89 | |
NM_009134.3 | NP_033160.2 | 20 - 21 | 3574 | 89 | ||
GenBank mRNA | AK157866.1 | - | 20 - 21 | 3574 | 89 | |
AY538273.1 | - | 19 - 20 | 3504 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mus musculus Probe spans exons |