Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011672.4 | NP_035802.3 | 10 - 11 | 887 | 103 | |
NR_028403.1 | 10 - 11 | 861 | 103 | |||
GenBank mRNA | AK010461.1 | - | 10 - 11 | 856 | 103 | |
AK088823.1 | - | 10 - 11 | 856 | 103 | ||
AK147138.1 | - | 10 - 11 | 870 | 103 | ||
AK169062.1 | - | 10 - 11 | 869 | 103 | ||
BC006630.1 | - | 10 - 11 | 844 | 3708320 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |