Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201607.2 | NP_963901.1 | 7 - 8 | 765 | 116 | |
GenBank mRNA | AK053713.1 | - | 5 - 6 | 524 | 116 | |
AK149514.1 | - | 7 - 8 | 715 | 116 | ||
BC030873.1 | - | 7 - 8 | 765 | 4223352 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |