Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012765.4 | NP_001012783.3 | 6 - 7 | 2270 | 60 | |
GenBank mRNA | AK147590.1 | - | 2373 | 60 | ||
AK147649.1 | - | 3030 | 60 | |||
AK160942.1 | - | 6 - 7 | 2270 | 60 | ||
BC035550.1 | - | 408 | 60 | |||
BC090846.1 | - | 6 - 7 | 2005 | 30363147 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |