Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018002.1 | NP_001018002.1 | 2 - 3 | 227 | 98 | |
NM_028610.2 | NP_082886.2 | 2 - 3 | 227 | 98 | ||
GenBank mRNA | AF490348.1 | - | 2 - 3 | 144 | 98 | |
AK010753.1 | - | 2 - 3 | 195 | 98 | ||
AK033197.1 | - | 2 - 3 | 228 | 98 | ||
BY731814.1 | - | 2 - 3 | 193 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |