Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007889.2 | NP_031915.2 | 13 - 14 | 1633 | 76 | |
GenBank mRNA | AK052151.1 | - | 13 - 14 | 1634 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |