Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600011085.mm.1 |
NM_183158.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011085.mm.1 |
NM_183158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011085.mm.1 |
TC1600001342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600011094.mm.1 |
NM_183158.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011094.mm.1 |
NM_183158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011094.mm.1 |
TC1600001342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600011086.mm.1 |
NM_183158.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011086.mm.1 |
NM_183158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011086.mm.1 |
TC1600001342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600011091.mm.1 |
NM_183158.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011091.mm.1 |
NM_183158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011091.mm.1 |
TC1600001342.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600011087.mm.1 |
NM_183158.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011087.mm.1 |
NM_183158.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600011087.mm.1 |
TC1600001342.mm.1 |
MTA TC |