Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144550.4 | NP_653133.3 | 1 - 2 | 108 | 58 | |
GenBank mRNA | AK048789.1 | - | 1 - 2 | 52 | 58 | |
AY099108.1 | - | 1 - 2 | 388 | 58 | ||
BC052381.1 | - | 1 - 2 | 62 | 5703283 | 58 | |
BC056960.1 | - | 1 - 2 | 62 | 5703283 | 58 | |
CX568108.1 | - | 1 - 2 | 108 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |