Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030711.4 | NP_109636.1 | 4 - 5 | 839 | 99 | |
GenBank mRNA | AB047552.1 | - | 3 - 4 | 827 | 99 | |
AF227511.1 | - | 3 - 4 | 815 | 99 | ||
AK029841.1 | - | 4 - 5 | 838 | 99 | ||
AK030329.1 | - | 4 - 5 | 840 | 99 | ||
AK036559.1 | - | 840 | 99 | |||
AK081181.1 | - | 4 - 5 | 866 | 99 | ||
AK163954.1 | - | 4 - 5 | 859 | 99 | ||
BC046610.1 | - | 4 - 5 | 845 | 6397585 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |