Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 2 - 3 | 221 | 73 | |
GenBank mRNA | AK040572.1 | - | 2 - 3 | 181 | 73 | |
AK041690.1 | - | 2 - 3 | 188 | 73 | ||
AK169696.1 | - | 2 - 3 | 176 | 73 | ||
BC027561.1 | - | 2 - 3 | 207 | 73 | ||
BY227020.1 | - | 2 - 3 | 221 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |