Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010918.2 | NP_035048.3 | 6 - 7 | 516 | 170 | |
GenBank mRNA | AK035582.1 | - | 5 - 6 | 417 | 170 | |
AK144214.1 | - | 6 - 7 | 427 | 170 | ||
L04289.2 | - | 6 - 7 | 521 | 170 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |