Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vill | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164567.1 | NP_001158039.1 | 12 - 13 | 1379 | 65 | |
NM_011700.2 | NP_035830.2 | 12 - 13 | 1379 | 65 | ||
GenBank mRNA | AJ344341.1 | - | 12 - 13 | 1367 | 65 | |
AK028229.1 | - | 12 - 13 | 1380 | 65 | ||
AK166823.1 | - | 11 - 12 | 1611 | 65 | ||
BC021808.1 | - | 12 - 13 | 1355 | 3991439 | 65 | |
BC022664.1 | - | 3 - 4 | 308 | 4223317 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |