Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175438.4 | NP_780647.3 | 12 - 13 | 1582 | 100 | |
GenBank mRNA | BC060650.1 | - | 11 - 12 | 2972 | 30354228 | 100 |
CD354968.1 | - | 2 - 3 | 214 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |