Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009951.1 | NP_001009951.1 | 4 - 5 | 458 | 146 | |
GenBank mRNA | AK220415.1 | - | 3 - 4 | 414 | 146 | |
BC088983.1 | - | 4 - 5 | 458 | 5700628 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |