Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004062.2 | NP_001004062.1 | 9 - 10 | 1027 | 64 | |
GenBank mRNA | AK129173.1 | - | 9 - 10 | 1024 | 64 | |
BC056137.1 | - | 8 - 9 | 5723 | 64 | ||
BC068139.1 | - | 9 - 10 | 1033 | 64 | ||
BC080308.1 | - | 9 - 10 | 1027 | 5719562 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |