Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040692.3 | NP_001035782.1 | 7 - 8 | 1066 | 81 | |
NM_001136087.2 | NP_001129559.1 | 7 - 8 | 1066 | 81 | ||
NM_001168643.1 | NP_001162114.1 | 7 - 8 | 1066 | 81 | ||
NM_001168644.1 | NP_001162115.1 | 7 - 8 | 1066 | 81 | ||
NM_001168645.1 | NP_001162116.1 | 7 - 8 | 1066 | 81 | ||
NM_001168646.1 | NP_001162117.1 | 7 - 8 | 1066 | 81 | ||
GenBank mRNA | AF075262.1 | - | 7 - 8 | 1043 | 81 | |
AF075263.1 | - | 7 - 8 | 1043 | 81 | ||
AF075264.1 | - | 7 - 8 | 1043 | 81 | ||
AF075265.1 | - | 7 - 8 | 1043 | 81 | ||
AF075266.1 | - | 7 - 8 | 1043 | 81 | ||
AF075267.1 | - | 7 - 8 | 1043 | 81 | ||
AK085258.1 | - | 7 - 8 | 1063 | 81 | ||
AK085473.1 | - | 7 - 8 | 1064 | 81 | ||
AK128907.1 | - | 7 - 8 | 1063 | 81 | ||
BC010748.1 | - | 7 - 8 | 1042 | 4235659 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |