Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301674.1 | NP_001288603.1 | 1 - 2 | 272 | 63 | |
GenBank mRNA | BC018226.1 | - | 1 - 2 | 271 | 3983850 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |