Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114087.2 | NP_001107559.1 | 5 - 6 | 494 | 120 | |
NM_001114088.2 | NP_001107560.1 | 5 - 6 | 494 | 120 | ||
GenBank mRNA | AK033091.1 | - | 486 | 120 | ||
AK077998.1 | - | 5 - 6 | 486 | 120 | ||
AK128987.1 | - | 5 - 6 | 482 | 120 | ||
AK142933.1 | - | 5 - 6 | 478 | 121 | ||
AK167476.1 | - | 5 - 6 | 486 | 120 | ||
BC052698.1 | - | 5 - 6 | 465 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |