Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030035.1 | NP_084311.1 | 14 - 15 | 8794 | 65 | |
GenBank mRNA | AK220188.1 | - | 2 - 3 | 4649 | 65 | |
BC040461.1 | - | 2 - 3 | 2114 | 65 | ||
BC060254.1 | - | 1 - 2 | 1218 | 65 | ||
BC150731.1 | - | 14 - 15 | 8771 | 9087642 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |