Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289915.1 | NP_001276844.1 | 3 - 4 | 456 | 71 | |
GenBank mRNA | AK172122.1 | - | 3 - 4 | 384 | 71 | |
BC072615.1 | - | 3 - 4 | 372 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |