Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172053.2 | NP_742050.2 | 18 - 19 | 3045 | 63 | |
GenBank mRNA | AK031314.1 | - | 18 - 19 | 3176 | 63 | |
AK173338.1 | - | 18 - 19 | 2981 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |