Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011571.3 | NP_035701.3 | 4 - 5 | 1675 | 76 | |
GenBank mRNA | AB003494.1 | - | 4 - 5 | 2013 | 76 | |
AK144302.1 | - | 4 - 5 | 1258 | 76 | ||
BC099699.1 | - | 4 - 5 | 1675 | 6821934 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |