Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phactr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005740.1 | NP_001005740.1 | 1 - 2 | 274 | 131 | |
GenBank mRNA | AK047654.1 | - | 3 - 4 | 432 | 131 | |
AK048208.1 | - | 1 - 2 | 170 | 131 | ||
AY993932.1 | - | 3 - 4 | 418 | 131 | ||
BC048407.1 | - | 1 - 2 | 312 | 131 | ||
BC061691.1 | - | 1 - 2 | 274 | 6307669 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |